3-Brom-5-chlor-2-hydroxybenzaldehyd
Lieferant: Thermo Scientific
Synonyme:
3-Bromo-5-chlorosalicylaldehyde
BTB03079.1
BTB03079.10
BTB03079.50
MAYBBTB03079.1EA
0
EUR
MAYBBTB03079.1
MAYBBTB03079.10
MAYBBTB03079.50
3-Brom-5-chlor-2-hydroxybenzaldehyd
3-Bromo-5-chloro-2-hydroxybenzaldehyde
Formel:
C₇H₄BrClO₂ Molecular Weight: 235,46 g/mol |
MDL:
MFCD00051690 CAS-Nummer: 19652-32-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...