4-Brom-2,6-dichloranilin ≥98%

Lieferant: Thermo Scientific
Gefahr

A14884.14 A14884.22
A14884.14EA 61.2 EUR
A14884.14 A14884.22
4-Brom-2,6-dichloranilin ≥98%
4-Brom-2,6-dichloranilin

Purity: 98% CAS No.: 697-88-1 Molecular Formula: C6H4BrCl2N Formula Weight: 240.92 Melting Point: 85-88° Beilstein Reference: 2089687

Formel: C₆H₄BrCl₂N
Molecular Weight: 240,91 g/mol
Siedepunkt: 290 °C (1013 hPa)
Schmelzpunkt: 85…88 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00025162
CAS-Nummer: 697-88-1
EINECS: 211-809-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR