4-Brom-2,6-dichloranilin
Lieferant: Thermo Scientific
|
Achtung
|
BTB03106.1
BTB03106.10
BTB03106.50
MAYBBTB03106.1EA
0
EUR
MAYBBTB03106.1
MAYBBTB03106.10
MAYBBTB03106.50
4-Brom-2,6-dichloranilin
4-Brom-2,6-dichloranilin
Formel:
C₆H₄BrCl₂N Molecular Weight: 240,91 g/mol Siedepunkt: 290 °C (1013 hPa) Schmelzpunkt: 90 °C |
MDL:
MFCD00025162 CAS-Nummer: 697-88-1 EINECS: 211-809-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...