3,5-Dibrom-1-fluorbenzol ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 1,3-Dibromo-5-fluorobenzene

A15078.14 A15078.22
A15078.14EA 121 EUR
A15078.14 A15078.22
3,5-Dibrom-1-fluorbenzol ≥98%
3,5-Dibrom-1-fluorbenzol

Purity: 98+% CAS No.: 1435-51-4 Molecular Formula: C6H3Br2F Formula Weight: 253.91 Flash Point: 110°(230°F) Refractive Index: 1.5770 Boiling Point: 204-206° Beilstein Reference: 2353462 MDL No.: MFCD00061119

Formel: C₆H₃Br₂F
Molecular Weight: 253,9 g/mol
Siedepunkt: 204…206 °C
Dichte: 2,018
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00061119
CAS-Nummer: 1435-51-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR