3,5-Dibrom-1-fluorbenzol
Lieferant: FLUOROCHEM
Synonyme:
1,3-Dibromo-5-fluorobenzene
004651-5G
004651-25G
004651-100G
FLUO004651-5GEA
0
EUR
FLUO004651-5G
FLUO004651-25G
FLUO004651-100G
3,5-Dibrom-1-fluorbenzol
3,5-Dibrom-1-fluorbenzol
Formel:
C₆H₃Br₂F Molecular Weight: 253.9 g/mol Siedepunkt: 204 Dichte: 2.01 g/cm³ (20 °C) |
MDL:
MFCD00061119 CAS-Nummer: 1435-51-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...