1,3-Dibromtetrafluorbenzol

Lieferant: FLUOROCHEM
Achtung

Synonyme: 1,3-Dibromo-2,4,5,6-tetrafluorobenzene

004667-25G 004667-5G
FLUO004667-25GEA 0 EUR
FLUO004667-25G FLUO004667-5G
1,3-Dibromtetrafluorbenzol
1,3-Dibromtetrafluorbenzol
Formel: C₆Br₂F₄
Molecular Weight: 307,87 g/mol
Siedepunkt: 85 °C (18 mmHg)
MDL: MFCD00031533
CAS-Nummer: 1559-87-1
EINECS: 216-326-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR