1-Brom-2,3,5,6-tetrafluorbenzol ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2,3,5,6-Tetrafluorbrombenzol
B24265.06
B24265.14
B24265.06EA
109
EUR
B24265.06
B24265.14
1-Brom-2,3,5,6-tetrafluorbenzol ≥99%
1-Brom-2,3,5,6-tetrafluorbenzol
Purity: 99%
CAS No.: 1559-88-2
Molecular Formula: C6HBrF4
Formula Weight: 228.98
Flash Point: 53°(129°F)
Refractive Index: 1.4690
Odour: Aromatic
Boiling Point: 142-144°
UN No.: 1993
MDL No.: MFCD00000308
Formel:
C₆HBrF₄ Molecular Weight: 228.97 g/mol Siedepunkt: 142…144 °C Dichte: 1.883 Flammpunkt: 53 °C (129 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000308 CAS-Nummer: 1559-88-2 EINECS: 216-327-9 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...