2-Brom-1,4-difluorbenzol ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2,5-Difluorobromobenzene, 1-Bromo-2,5-difluorobenzene
A12963.06
A12963.14
A12963.22
A12963.06EA
37.6
EUR
A12963.06
A12963.14
A12963.22
2-Brom-1,4-difluorbenzol ≥98%
2-Brom-1,4-difluorbenzol
Purity: 98%
CAS No.: 399-94-0
Molecular Formula: C6H3BrF2
Formula Weight: 192.99
Flash Point: 65°(149°F)
Refractive Index: 1.5080
Melting Point: -31°
Boiling Point: 73-75°/65mm
Beilstein Reference: 1680893
MDL No.: MFCD00000345
Formel:
C₆H₃BrF₂ Molecular Weight: 192,99 g/mol Siedepunkt: 73…75 °C (65 mmHg) Schmelzpunkt: –31 °C Dichte: 1,714 Flammpunkt: 65 °C (149 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000345 CAS-Nummer: 399-94-0 EINECS: 206-920-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...