1-Brom-2-fluorbenzol
Lieferant: FLUOROCHEM
Synonyme:
2-Bromofluorobenzene
001121-100G
001121-500G
001121-25G
FLUO001121-100GEA
136
EUR
FLUO001121-100G
FLUO001121-500G
FLUO001121-25G
1-Brom-2-fluorbenzol
1-Brom-2-fluorbenzol
Formel:
C₆H₄BrF Molecular Weight: 175 g/mol Siedepunkt: 155 Dichte: 1.61 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000282 CAS-Nummer: 1072-85-1 EINECS: 214-018-3 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...