1-Brom-3-fluorbenzol ≥99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-Bromofluorobenzene
A10858.14
A10858.18
A10858.22
A10858.30
A10858.09
A10858.14EA
47.1
EUR
A10858.14
A10858.18
A10858.22
A10858.30
A10858.09
1-Brom-3-fluorbenzol ≥99%
1-Brom-3-fluorbenzol
Purity: 99%
CAS No.: 1073-06-9
Molecular Formula: C6H4BrF
Formula Weight: 175.01
Flash Point: 46°(115°F)
Refractive Index: 1.5275
Odour: Aromatic
Melting Point: -8°
Boiling Point: 148-150°
Beilstein Reference: 1853969
UN No.: 1993
MDL No.: MFCD00000326
Formel:
C₆H₄BrF Molecular Weight: 175 g/mol Siedepunkt: 148…150 °C Schmelzpunkt: –8 °C Dichte: 1,594 Flammpunkt: 46 °C (115 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000326 CAS-Nummer: 1073-06-9 EINECS: 214-023-0 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...