Chlordimethylvinylsilan

Lieferant: abcr

Synonyme: Dimethylvinylchlorsilan, Vinyldimethylchlorsilan

AB109176.0100 AB109176.0025 AB109176.0250
ABCRAB109176.0100EA 0 EUR
ABCRAB109176.0100 ABCRAB109176.0025 ABCRAB109176.0250
Chlordimethylvinylsilan
Chlordimethylvinylsilan
Formel: C₄H₉ClSi
Molecular Weight: 120.65 g/mol
MDL: MFCD00018090
CAS-Nummer: 1719-58-0
EINECS: 217-007-1
UN: 2985
ADR: 3 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR