Bis(4-fluorphenyl)sulfon 99%
Lieferant: Thermo Fisher Scientific
Synonyme:
4,4'-Difluordiphenylsulfon, 4-Fluorphenylsulfon
119620250
119621000
ACRO119620250EA
94.8
EUR
ACRO119620250
ACRO119621000
Bis(4-fluorphenyl)sulfon 99%
Bis(4-fluorphenyl)sulfon
Formel:
C₁₂H₈F₂O₂S Molecular Weight: 254.26 g/mol Schmelzpunkt: 97…100 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000350 CAS-Nummer: 383-29-9 EINECS: 206-847-4 |
Spezifikation Testergebnisse
Appearance | White to brown Powder or crystals |
Infrared spectrum | Conforms |
Melting point | 97 to 100 °C |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...