Tetramethylensulfoxid 97%
Lieferant: Thermo Scientific
Synonyme:
Tetrahydrothiophen-1-oxid, Thiacyclopentan-1-oxid
138440250
138441000
138440050
ACRO138440250EA
73.8
EUR
ACRO138440250
ACRO138441000
ACRO138440050
Tetramethylensulfoxid 97%
Tetramethylensulfoxid
Formel:
C₄H₈OS Molecular Weight: 104,17 g/mol Flammpunkt: >112 °C |
MDL:
MFCD00005477 CAS-Nummer: 1600-44-8 EINECS: 216-493-2 |
Spezifikation Testergebnisse
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5190 to 1.5230 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...