4-Bromphenylhydraziniumchlorid 99%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 1-(4-Bromphenyl)hydrazin Hydrochlorid, 4-Bromphenylhydrazinhydrochlorid, 4-Bromphenylhydrazin Hydrochlorid

162310100 162310500
ACRO162310100EA 84.1 EUR
ACRO162310100 ACRO162310500
4-Bromphenylhydraziniumchlorid 99%
4-Bromphenylhydraziniumchlorid
Formel: C₆H₈BrClN₂
Molecular Weight: 223.50 g/mol
Schmelzpunkt: 210…215 °C
MDL: MFCD00012941
CAS-Nummer: 622-88-8
UN: 3261

Bestellen

Spezifikation Testergebnisse

Appearance Beige to grey to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 210 to 215 °C (Dec)
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR