3-Fluorphenylhydraziniumchlorid 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
1-(3-Fluorphenyl)hydrazin Hydrochlorid, 3-Fluorphenylhydrazin Hydrochlorid, 3-Fluorphenylhydrazinhydrochlorid
167370050
167370100
ACRO167370050EA
46.1
EUR
ACRO167370050
ACRO167370100
3-Fluorphenylhydraziniumchlorid 97%
3-Fluorphenylhydraziniumchlorid
Formel:
C₆H₈ClFN₂ Molecular Weight: 162.59 g/mol Schmelzpunkt: 268 °C (Zers.) |
MDL:
MFCD00012934 CAS-Nummer: 2924-16-5 EINECS: 220-886-4 UN: 2811 |
Spezifikation Testergebnisse
Appearance | White to yellow to light brown Powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...