Kaliumosmat(VI) Dihydrat ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Kaliumosmium(VI)oxid Dihydrat, Kaliumosmat Dihydrat, Dikaliumosmat Dihydrat

12647.01 12647.03
12647.01EA 66.4 EUR
12647.01 12647.03
Kaliumosmat(VI) Dihydrat ≥98%
Kaliumosmat(VI) Dihydrat

Purity: 98+% CAS No.: 10022-66-9 Molecular Formula: K2OsO4.2H2O Formula Weight: 368.43 (332.40anhy) Physical Form: Powder Sensitivity: Hygroscopic Appearance: Purple Solubility: Soluble in water. Slowly decomposes in aqueous solution forming the tetroxide. Insoluble in alcohol, ether UN No.: 3288 MDL No.: MFCD00149919

Formel: K₂O₄Os·2H₂O
Molecular Weight: 368,45 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00149919
CAS-Nummer: 10022-66-9
EINECS: 243-247-1
UN: 3288
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR