2,4-Dichlorphenylhydraziniumchlorid ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
1-(2,4-Dichlorphenyl)hydrazin Hydrochlorid, 2,4-Dichlorphenylhydrazinhydrochlorid, 2,4-Dichlorphenylhydrazin Hydrochlorid
A10943.09
A10943.18
A10943.30
A10943.09EA
67
EUR
A10943.09
A10943.18
A10943.30
2,4-Dichlorphenylhydraziniumchlorid ≥98%
2,4-Dichlorphenylhydraziniumchlorid
Purity: 98%
CAS No.: 5446-18-4
Molecular Formula: C6H6Cl2N2.HCl
Formula Weight: 213.49
Melting Point: ca 212° dec.
Beilstein Reference: 3708828
Formel:
C₆H₇Cl₃N₂ Molecular Weight: 213,49 g/mol Schmelzpunkt: ∼ 212 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012929 CAS-Nummer: 5446-18-4 EINECS: 226-659-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...