Methyl-3,5-dimethoxybenzoat ≥98%

Lieferant: Thermo Scientific

Synonyme: 3,5-Dimethoxybenzoesäuremethylester

A11486.09 A11486.18
A11486.09EA 34 EUR
A11486.09 A11486.18
Methyl-3,5-dimethoxybenzoat ≥98%
Methyl-3,5-dimethoxybenzoat

Purity: 98% CAS No.: 2150-37-0 Molecular Formula: C10H12O4 Formula Weight: 196.20 Flash Point: 110°(230°F) Melting Point: 40-43° Boiling Point: 298° Beilstein Reference: 2693126 MDL No.: MFCD00008432

Formel: C₁₀H₁₂O₄
Molecular Weight: 196,2 g/mol
Siedepunkt: 298 °C
Schmelzpunkt: 40…43 °C
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008432
CAS-Nummer: 2150-37-0
EINECS: 218-423-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR