2,6-Dichlorphenylhydraziniumchlorid ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 2,6-Dichlorphenylhydrazinhydrochlorid, 2,6-Dichlorphenylhydrazin Hydrochlorid, 1-(2,6-Dichlorphenyl)hydrazin Hydrochlorid

A11968.03 A11968.06
A11968.03EA 32.3 EUR
A11968.03 A11968.06
2,6-Dichlorphenylhydraziniumchlorid ≥98%
2,6-Dichlorphenylhydraziniumchlorid

Purity: 98+% CAS No.: 50709-36-9 Molecular Formula: C6H6Cl2N2.HCl Formula Weight: 213.50 Melting Point: ca 225° dec. Beilstein Reference: 4569738

Formel: C₆H₇Cl₃N₂
Molecular Weight: 213,49 g/mol
Schmelzpunkt: ∼ 225 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00012930
CAS-Nummer: 50709-36-9
EINECS: 256-730-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR