(3-Fluor-4-hydroxyphenyl)essigsäure ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-Fluoro-4-hydroxyphenylacetic acid

A12114.06 A12114.03
A12114.06EA 91 EUR
A12114.06 A12114.03
(3-Fluor-4-hydroxyphenyl)essigsäure ≥98%
(3-Fluor-4-hydroxyphenyl)essigsäure

Purity: 98% CAS No.: 458-09-3 Molecular Formula: C8H7FO3 Formula Weight: 170.14 Melting Point: 130-133° MDL No.: MFCD00004348

Formel: C₈H₇FO₃
Molecular Weight: 170,14 g/mol
Schmelzpunkt: 130…133 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004348
CAS-Nummer: 458-09-3
EINECS: 207-275-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR