Methyl-4,5-dimethoxy-2-nitrobenzoat ≥97%
Lieferant: Thermo Fisher Scientific
Synonyme: Methyl-6-nitroveratrumat, 4,5-Dimethoxy-2-nitrobenzoesäuremethylester
A12836.18 A12836.30 A12836.36
A12836.18EA 43.9 EUR
A12836.18 A12836.30 A12836.36
Methyl-4,5-dimethoxy-2-nitrobenzoat ≥97%
Methyl-4,5-dimethoxy-2-nitrobenzoat
Purity: 97% CAS No.: 26791-93-5 Molecular Formula: C10H11NO6 Formula Weight: 241.20 Melting Point: 142-144° Beilstein Reference: 2381747
Formel: C₁₀H₁₁NO₆
Molecular Weight: 241.2 g/mol
Schmelzpunkt: 142…144 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007239
CAS-Nummer: 26791-93-5
EINECS: 248-008-5