Ethyl-3,4,5-trimethoxybenzoat ≥98%
Lieferant: Thermo Scientific
Synonyme:
3,4,5-Trimethoxybenzoesäureethylester
A13199.06
A13199.22
A13199.06EA
18.2
EUR
A13199.06
A13199.22
Ethyl-3,4,5-trimethoxybenzoat ≥98%
Ethyl-3,4,5-trimethoxybenzoat
Purity: 98%
CAS No.: 6178-44-5
Molecular Formula: C12H16O5
Formula Weight: 240.26
MDL No.: MFCD00017525
Formel:
C₁₂H₁₆O₅ Molecular Weight: 240,26 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00017525 CAS-Nummer: 6178-44-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...