Methyl-4-amino-2-methoxybenzoat 98%
Lieferant: Thermo Scientific
Synonyme:
4-Amino-2-methoxybenzoesäuremethylester
A13239.06
A13239.14
A13239.06EA
23.2
EUR
A13239.06
A13239.14
Methyl-4-amino-2-methoxybenzoat 98%
Methyl-4-amino-2-methoxybenzoat
Purity: 98%
CAS No.: 27492-84-8
Molecular Formula: C9H11NO3
Formula Weight: 181.19
Melting Point: 154-157°
MDL No.: MFCD00017202
Formel:
C₉H₁₁NO₃ Molecular Weight: 181,19 g/mol Schmelzpunkt: 154…157 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00017202 CAS-Nummer: 27492-84-8 EINECS: 248-494-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...