Sulfolan ≥99%

Lieferant: Thermo Scientific
Achtung

Synonyme: Thiacyclopentan-1,1-dioxid, Tetrahydrothiophen-1,1-dioxid, Tetramethylensulfon

A13466.0B A13466.30 A13466.0I
A13466.0BEA 122 EUR
A13466.0B A13466.30 A13466.0I
Sulfolan ≥99%
Sulfolan

Purity: 99% CAS No.: 126-33-0 Molecular Formula: C4H8O2S Formula Weight: 120.17 Flash Point: 166°(330°F) Refractive Index: 1.4825 Odour: Weak, characteristic Melting Point: 25-28° Boiling Point: 285° Beilstein Reference: 107765 Merck Reference: 14,8955 MDL No.: MFCD00005484

Formel: C₄H₈O₂S
Molecular Weight: 120,17 g/mol
Siedepunkt: 285 °C
Schmelzpunkt: 25…28 °C
Dichte: 1,261
Flammpunkt: 166 °C (330 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00005484
CAS-Nummer: 126-33-0
EINECS: 204-783-1
Merck Index: 14,8955

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR