m-Tolylhydraziniumchlorid ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3-Methylphenylhydrazin Hydrochlorid, 1-(3-Methylphenyl)hydrazin Hydrochlorid, 3-Tolylhydrazin Hydrochlorid, 3-Methylphenylhydraziniumchlorid, 1-(3-Tolyl)hydrazin Hydrochlorid, 3-Tolylhydrazinhydrochlorid, 3-Methylphenylhydrazinhydrochlorid
A13667.06
A13667.14
A13667.06EA
64.4
EUR
A13667.06
A13667.14
m-Tolylhydraziniumchlorid ≥98%
m-Tolylhydraziniumchlorid
Purity: 98%
CAS No.: 637-04-7
Molecular Formula: C7H10N2.HCl
Formula Weight: 158.63
Melting Point: ca 193° dec.
Beilstein Reference: 3563995
UN No.: 2811
MDL No.: MFCD00012932
Formel:
C₇H₁₀N₂·HCl Molecular Weight: 158.63 g/mol Schmelzpunkt: ∼ 193 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012932 CAS-Nummer: 637-04-7 EINECS: 211-276-9 UN: 2811 ADR: 6.1 III |