2-Fluorphenylhydraziniumchlorid ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 2-Fluorphenylhydrazin Hydrochlorid, 1-(2-Fluorphenyl)hydrazin Hydrochlorid, 2-Fluorphenylhydrazinhydrochlorid

A13716.03 A13716.06
A13716.03EA 27.4 EUR
A13716.03 A13716.06
2-Fluorphenylhydraziniumchlorid ≥98%
2-Fluorphenylhydraziniumchlorid

Purity: 98% CAS No.: 2924-15-4 Molecular Formula: C6H7FN2.HCl Formula Weight: 162.60 Melting Point: ca 205° dec. Beilstein Reference: 3627985

Formel: C₆H₈ClFN₂
Molecular Weight: 162.59 g/mol
Schmelzpunkt: ∼ 205 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00012927
CAS-Nummer: 2924-15-4
EINECS: 220-885-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR