4-Methoxybenzaldehyddimethylacetal ≥98%
Lieferant: Thermo Scientific
Synonyme:
1-(Dimethoxymethyl)-4-methoxybenzol, 4-Methoxy-α,α-dimethoxytoluol, p-Anisaldehyddimethylacetal, p-(Dimethoxymethyl)anisol
A15793.14
A15793.18
A15793.30
A15793.36
A15793.14EA
43.9
EUR
A15793.14
A15793.18
A15793.30
A15793.36
4-Methoxybenzaldehyddimethylacetal ≥98%
4-Methoxybenzaldehyddimethylacetal
Purity: 98%
CAS No.: 2186-92-7
Molecular Formula: C10H14O3
Formula Weight: 182.22
Flash Point: 97°(206°F)
Refractive Index: 1.5055
Odour: Pleasant
Boiling Point: 253°
Beilstein Reference: 2048930
MDL No.: MFCD00036507
Formel:
C₁₀H₁₄O₃ Molecular Weight: 182,22 g/mol Siedepunkt: 253 °C Dichte: 1,068 Flammpunkt: 97 °C (206 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00036507 CAS-Nummer: 2186-92-7 EINECS: 218-577-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...