Nitrosiumtetrafluoroborat ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Nitrosyltetrafluoroborat
A15806.04
A15806.09
A15806.18
A15806.04EA
30
EUR
A15806.04
A15806.09
A15806.18
Nitrosiumtetrafluoroborat ≥98%
Nitrosium tetrafluoroborate
Purity: 98%
CAS No.: 14635-75-7
Molecular Formula: NOBF4
Formula Weight: 116.82
Storage Temperature: 2°C to 8°C
Odour: Irritating
Boiling Point: 250°/0.01mm subl.
Merck Reference: 14,6649
UN No.: 3260
MDL No.: MFCD00011433
Formel:
BF₄NO Molecular Weight: 116,81 g/mol Siedepunkt: 250 °C (0,01 mmHg) (Subl.) Schmelzpunkt: 250 °C (Subl.) Dichte: 2,185 |
MDL:
MFCD00011433 CAS-Nummer: 14635-75-7 EINECS: 238-679-2 UN: 3260 ADR: 8,II Merck Index: 14,6649 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...