Bis(4-hydroxyphenyl)sulfon ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4,4'-Dihydroxydiphenylsulfon, 4-Hydroxyphenylsulfon, 4,4'-Sulfonyldiphenol

A17342.0B A17342.30
A17342.0BEA 132 EUR
A17342.0B A17342.30
Bis(4-hydroxyphenyl)sulfon ≥99%
Bis(4-hydroxyphenyl)sulfon

Purity: 99% CAS No.: 80-09-1 Molecular Formula: C12H10O4S Formula Weight: 250.28 Melting Point: 245-250° Beilstein Reference: 2052954

Formel: C₁₂H₁₀O₄S
Schmelzpunkt: 245…250 °C
Dichte: 1,366
Lagertemperatur: Raumtemperatur
MDL: MFCD00002350
CAS-Nummer: 80-09-1
EINECS: 201-250-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR