p-Toluolsulfonylisocyanat ≥95%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
4-Toluolsulfonylisocyanat, Tosylisocyanat
A17492.22
A17492.36
A17492.22EA
42.7
EUR
A17492.22
A17492.36
p-Toluolsulfonylisocyanat ≥95%
p-Toluolsulfonylisocyanat
Purity: 95%
CAS No.: 4083-64-1
Molecular Formula: C8H7NO3S
Formula Weight: 197.21
Flash Point: 145°(293°F)
Refractive Index: 1.5350
Odour: Irritating
Melting Point: 5°
Boiling Point: 144°/10mm
Beilstein Reference: 391287
UN No.: 2206
MDL No.: MFCD00002030
Formel:
C₈H₇NO₃S Molecular Weight: 197,21 g/mol Siedepunkt: 144 °C (10 mmHg) Schmelzpunkt: 5 °C Dichte: 1,295 Flammpunkt: 145 °C (293 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002030 CAS-Nummer: 4083-64-1 EINECS: 223-810-8 UN: 2206 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...