Methyl-o-anisat ≥98%
Lieferant: Thermo Scientific
Synonyme:
Methyl-2-methoxybenzoat, 2-Methoxybenzoesäuremethylester, o-Anissäuremethylester, 2-Anissäuremethylester
A17703.14
A17703.22
A17703.14EA
30.5
EUR
A17703.14
A17703.22
Methyl-o-anisat ≥98%
Methyl-o-anisat
Purity: 98+%
CAS No.: 606-45-1
Molecular Formula: C9H10O3
Formula Weight: 166.18
Flash Point:
Formel:
C₉H₁₀O₃ Molecular Weight: 166,18 g/mol Siedepunkt: 246…248 °C Dichte: 1,160 Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008423 CAS-Nummer: 606-45-1 EINECS: 210-118-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...