4-(2-Methyl-2-butanyl)phenol ≥99%

Lieferant: Thermo Scientific
Gefahr

Synonyme: p-(1,1-Dimethylpropyl)phenol, 4-(2-Methyl-2-butyl)phenol, 4-tert-Pentylphenol

B20939.18 B20939.30
B20939.18EA 31.9 EUR
B20939.18 B20939.30
4-(2-Methyl-2-butanyl)phenol ≥99%
4-tert-Amylphenol

Purity: 99% CAS No.: 80-46-6 Molecular Formula: C11H16O Formula Weight: 164.25 Melting Point: 88-89° Boiling Point: 254-256° Merck Reference: 14,7142 UN No.: 2430 MDL No.: MFCD00002369

Formel: C₁₁H₁₆O
Molecular Weight: 164,25 g/mol
Siedepunkt: 254…256 °C
Schmelzpunkt: 88…89 °C
Dichte: 0,962
Lagertemperatur: Raumtemperatur
MDL: MFCD00002369
CAS-Nummer: 80-46-6
EINECS: 201-280-9
UN: 2430
ADR: 8,III
Merck Index: 14,7142

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR