(4-Hydroxyphenyl)essigsäuremethylester ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Methyl-(4-hydroxyphenyl)acetat

B22533.14 B22533.22 B22533.36
B22533.14EA 46.2 EUR
B22533.14 B22533.22 B22533.36
(4-Hydroxyphenyl)essigsäuremethylester ≥98%
(4-Hydroxyphenyl)essigsäuremethylester

Purity: 98+% CAS No.: 14199-15-6 Molecular Formula: C9H10O3 Formula Weight: 166.18 Melting Point: 55-58° Boiling Point: 163°/5mm MDL No.: MFCD00002387

Formel: C₉H₁₀O₃
Molecular Weight: 166,18 g/mol
Siedepunkt: 163 °C (5 mmHg)
Schmelzpunkt: 55…58 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002387
CAS-Nummer: 14199-15-6
EINECS: 238-050-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR