Methyl-3,4-dimethoxybenzoat ≥98%
Lieferant: Thermo Fisher Scientific
Synonyme:
3,4-Dimethoxybenzoesäuremethylester, Methylveratrumat
B23568.18
AB131937.0050
B23568.18EA
50.9
EUR
B23568.18
ABCRAB131937.0050
Methyl-3,4-dimethoxybenzoat ≥98%
Methyl-3,4-dimethoxybenzoat
Purity: 98+%
CAS No.: 2150-38-1
Molecular Formula: C10H12O4
Formula Weight: 196.20
Melting Point: 57-61°
Boiling Point: 283°
MDL No.: MFCD00008430
Formel:
C₁₀H₁₂O₄ Molecular Weight: 196.2 g/mol Siedepunkt: 283 °C Schmelzpunkt: 57…61 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008430 CAS-Nummer: 2150-38-1 EINECS: 218-424-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...