2-(1-Piperidinyl)ethanol ≥99%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 1-(2-Hydroxyethyl)piperidin

B23816.22 B23816.36
B23816.22EA 45.3 EUR
B23816.22 B23816.36
2-(1-Piperidinyl)ethanol ≥99%
2-Piperidinoethanol

Purity: 99% CAS No.: 3040-44-6 Molecular Formula: C7H15NO Formula Weight: 129.20 Flash Point: 83°(181°F) Refractive Index: 1.4804 Odour: Amine-like pH: 12.4 Melting Point: 16° Boiling Point: 199-202° Beilstein Reference: 103390 UN No.: 2735 MDL No.: MFCD00006512

Formel: C₇H₁₅NO
Molecular Weight: 129,2 g/mol
Siedepunkt: 199…202 °C
Schmelzpunkt: 16 °C
Dichte: 0,977
Flammpunkt: 83 °C (181 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00006512
CAS-Nummer: 3040-44-6
EINECS: 221-244-6
UN: 2735
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR