2-Bromphenylhydraziniumchlorid 95%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 2-Bromphenylhydrazin Hydrochlorid, 1-(2-Bromphenyl)hydrazin Hydrochlorid, 2-Bromphenylhydrazinhydrochlorid

B23966.06 B23966.14 B23966.22
B23966.06EA 63.4 EUR
B23966.06 B23966.14 B23966.22
2-Bromphenylhydraziniumchlorid 95%
2-Bromphenylhydraziniumchlorid

Purity: 95% CAS No.: 50709-33-6 Molecular Formula: C6H7BrN2.HCl Formula Weight: 223.51 Melting Point: ca 188° dec. MDL No.: MFCD00012926

Formel: C₆H₈BrClN₂
Molecular Weight: 223,5 g/mol
Schmelzpunkt: ∼ 188 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00012926
CAS-Nummer: 50709-33-6
EINECS: 256-728-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR