1H,1H-Pentafluorpropylmethacrylat ≥97% stabilisiert
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Methylpropensäure(2,2,2,3,3-pentafluorpropyl)ester, 1H,1H-Pentafluorpropylmethacrylat, 1H,1H-Perfluorpropylmethacrylat, Methacrylsäure(2,2,2,3,3-pentafluorpropyl)ester
B24340.06
B24340.14
B24340.06EA
62.3
EUR
B24340.06
B24340.14
1H,1H-Pentafluorpropylmethacrylat ≥97% stabilisiert
1H,1H-Pentafluorpropylmethacrylat
Stabilized
Purity: 97%
CAS No.: 45115-53-5
Molecular Formula: C7H7F5O2
Formula Weight: 218.12
Storage Temperature: 2°C to 8°C
Flash Point: 29°(84°F)
Refractive Index: 1.3480
Boiling Point: 54-55°/100mm
UN No.: 3272
MDL No.: MFCD00039256
Formel:
C₇H₇F₅O₂ Molecular Weight: 218,12 g/mol Siedepunkt: 54…55 °C (100 mmHg) Dichte: 1,279 Flammpunkt: 29 °C (84 °F) |
MDL:
MFCD00039256 CAS-Nummer: 45115-53-5 EINECS: 256-191-8 UN: 3272 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...