4-Brom-2-fluor-6-nitroanisol ≥97%

Lieferant: Thermo Scientific
Achtung

H30220.06 H30220.14
H30220.06EA 81.6 EUR
H30220.06 H30220.14
4-Brom-2-fluor-6-nitroanisol ≥97%
4-Brom-2-fluor-6-nitroanisol

Purity: 97% CAS No.: 74266-66-3 Molecular Formula: C7H5BrFNO3 Formula Weight: 250.02 Melting Point: 53-56° MDL No.: MFCD00013374

Formel: C₇H₅BrFNO₃
Molecular Weight: 250,02 g/mol
Schmelzpunkt: 53…56 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00013374
CAS-Nummer: 74266-66-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR