3-Brom-5-methoxybenzoesäure ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
H32635.03
H32635.06
H32635.03EA
35.2
EUR
H32635.03
H32635.06
3-Brom-5-methoxybenzoesäure ≥98%
3-Bromo-5-methoxybenzoic acid
Purity: 98%
CAS No.: 157893-14-6
Molecular Formula: C8H7BrO3
Formula Weight: 231.05
MDL No.: MFCD00266858
Formel:
C₈H₇BrO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00266858 CAS-Nummer: 157893-14-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...