3-Fluorphenylhydraziniumchlorid ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 1-(3-Fluorphenyl)hydrazin Hydrochlorid, 3-Fluorphenylhydrazin Hydrochlorid, 3-Fluorphenylhydrazinhydrochlorid

L07418.06 L07418.14
L07418.06EA 67.7 EUR
L07418.06 L07418.14
3-Fluorphenylhydraziniumchlorid ≥98%
3-Fluorphenylhydraziniumchlorid

Purity: 98% CAS No.: 2924-16-5 Molecular Formula: C6H7FN2.HCl Formula Weight: 162.60 Melting Point: ca 268° dec. Beilstein Reference: 3627729

Formel: C₆H₈ClFN₂
Molecular Weight: 162,59 g/mol
Schmelzpunkt: ∼ 268 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00012934
CAS-Nummer: 2924-16-5
EINECS: 220-886-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR