3-(Trifluormethyl)phenylhydrazin 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3-Hydrazinobenzotrifluorid, 1-(3-(Trifluormethyl)phenyl)hydrazin

L11996.03 L11996.06
L11996.03EA 55.5 EUR
L11996.03 L11996.06
3-(Trifluormethyl)phenylhydrazin 95%
3-(Trifluormethyl)phenylhydrazin

Purity: 95% CAS No.: 368-78-5 Molecular Formula: C7H7F3N2 Formula Weight: 176.14 Flash Point: 107°(224°F) Refractive Index: 1.5040 Boiling Point: 80-83°/9mm Beilstein Reference: 640075 MDL No.: MFCD00025093

Formel: C₇H₇F₃N₂
Molecular Weight: 176.14
Siedepunkt: 80…83 °C (9 mmHg)
Dichte: 1.348
Flammpunkt: 107 °C (224 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00025093
CAS-Nummer: 368-78-5
EINECS: 206-713-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR