3-Hydroxy-2-nitrobenzoesäure ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
L12841.09
L12841.04
L12841.09EA
459
EUR
L12841.09
L12841.04
3-Hydroxy-2-nitrobenzoesäure ≥98%
3-Hydroxy-2-nitrobenzoic acid
Purity: 98+%
CAS No.: 602-00-6
Molecular Formula: C7H5NO5
Formula Weight: 183.12
Melting Point: 177-181°
Beilstein Reference: 2372248
Formel:
C₇H₅NO₅ Molecular Weight: 183,12 g/mol Schmelzpunkt: 177…181 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00017003 CAS-Nummer: 602-00-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...