4'-Brom-3'-nitroacetophenon

Lieferant: CHEMPUR

Synonyme: 1-(4-Brom-3-nitrophenyl)ethan-1-on

BTB00581.250 BTB00581.1
CHMPBTB00581.250EA 0 EUR
CHMPBTB00581.250 MAYBBTB00581.1
4'-Brom-3'-nitroacetophenon
4'-Brom-3'-nitroacetophenon
Formel: C₈H₆BrNO₃
Schmelzpunkt: 120 °C
MDL: MFCD00016985
CAS-Nummer: 18640-58-9
EINECS: 242-469-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR