Methyl-2-amino-3,4,5-trimethoxybenzoat

Lieferant: Thermo Scientific

Synonyme: 2-Amino-3,4,5-trimethoxybenzoesäuremethylester, Methyl-3,4,5-trimethoxyanthranilat

BTB05442.1 BTB05442.10 BTB05442.50
MAYBBTB05442.1EA 0 EUR
MAYBBTB05442.1 MAYBBTB05442.10 MAYBBTB05442.50
Methyl-2-amino-3,4,5-trimethoxybenzoat
Methyl-2-amino-3,4,5-trimethoxybenzoat
Formel: C₁₁H₁₅NO₅
Molecular Weight: 241,24 g/mol
MDL: MFCD00008426
CAS-Nummer: 5035-82-5
EINECS: 225-728-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR