4-Chlorphenylhydraziniumchlorid
Lieferant: Thermo Scientific
Synonyme:
4-Chlorphenylhydrazinhydrochlorid, 4-Chlorphenylhydrazin Hydrochlorid, 1-(4-Chlorphenyl)hydrazin Hydrochlorid
BTB07482.1
BTB07482.10
BTB07482.50
MAYBBTB07482.1EA
0
EUR
MAYBBTB07482.1
MAYBBTB07482.10
MAYBBTB07482.50
4-Chlorphenylhydraziniumchlorid
4-Chlorphenylhydraziniumchlorid
Formel:
C₆H₈Cl₂N₂ Molecular Weight: 179.05 g/mol |
MDL:
MFCD00012943 CAS-Nummer: 1073-70-7 EINECS: 214-030-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...