2-Chlorphenylhydraziniumchlorid

Lieferant: Thermo Scientific

Synonyme: 2-Chlorphenylhydrazin Hydrochlorid, 2-Chlorphenylhydrazinhydrochlorid, 1-(2-Chlorphenyl)hydrazin Hydrochlorid

BTB07527.1 BTB07527.50
MAYBBTB07527.1EA 0 EUR
MAYBBTB07527.1 MAYBBTB07527.50
2-Chlorphenylhydraziniumchlorid
2-Chlorphenylhydraziniumchlorid
Formel: C₆H₈Cl₂N₂
Molecular Weight: 179.05 g/mol
MDL: MFCD00012928
CAS-Nummer: 41052-75-9
EINECS: 255-194-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR