m-Tolylhydraziniumchlorid

Lieferant: Thermo Scientific

Synonyme: 3-Methylphenylhydrazin Hydrochlorid, 1-(3-Methylphenyl)hydrazin Hydrochlorid, 3-Tolylhydrazin Hydrochlorid, 3-Methylphenylhydraziniumchlorid, 1-(3-Tolyl)hydrazin Hydrochlorid, 3-Tolylhydrazinhydrochlorid, 3-Methylphenylhydrazinhydrochlorid

BTB07668.1 BTB07668.10 BTB07668.50
MAYBBTB07668.1EA 0 EUR
MAYBBTB07668.1 MAYBBTB07668.10 MAYBBTB07668.50
m-Tolylhydraziniumchlorid
m-Tolylhydraziniumchlorid
Formel: C₇H₁₀N₂·HCl
Molecular Weight: 158.63 g/mol
MDL: MFCD00012932
CAS-Nummer: 637-04-7
EINECS: 211-276-9
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR