3,4-Dichlorphenylhydraziniumchlorid

Lieferant: Thermo Scientific

Synonyme: 3,4-Dichlorphenylhydrazinhydrochlorid, 1-(3,4-Dichlorphenyl)hydrazin Hydrochlorid, 3,4-Dichlorphenylhydrazin Hydrochlorid

BTB08664.1 BTB08664EA BTB08664.50
MAYBBTB08664.1EA 0 EUR
MAYBBTB08664.1 MAYBBTB08664EA MAYBBTB08664.50
3,4-Dichlorphenylhydraziniumchlorid
3,4-Dichlorphenylhydraziniumchlorid
Formel: C₆H₇Cl₃N₂
Molecular Weight: 213,49 g/mol
MDL: MFCD00012937
CAS-Nummer: 19763-90-7
EINECS: 243-278-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR