2,3-Dichlorphenylhydraziniumchlorid
Lieferant: Thermo Scientific
Synonyme: 2,3-Dichlorphenylhydrazinhydrochlorid, 2,3-Dichlorphenylhydrazin Hydrochlorid, 1-(2,3-Dichlorphenyl)hydrazin Hydrochlorid
BTB08666.1 BTB08666.10 BTB08666.50
MAYBBTB08666.1EA 0 EUR
MAYBBTB08666.1 MAYBBTB08666.10 MAYBBTB08666.50
2,3-Dichlorphenylhydraziniumchlorid
2,3-Dichlorphenylhydraziniumchlorid
Formel: C₆H₇Cl₃N₂
Molecular Weight: 213,49 g/mol
MDL: MFCD00052265
CAS-Nummer: 21938-47-6