2,5-Difluorphenylhydraziniumchlorid

Lieferant: CHEMPUR

Synonyme: 1-(2,5-Difluorphenyl)hydrazin Hydrochlorid, 2,5-Difluorphenylhydrazin Hydrochlorid, 2,5-Difluorphenylhydrazinhydrochlorid

BTB08668.250 BTB08668.1
CHMPBTB08668.250EA 0 EUR
CHMPBTB08668.250 MAYBBTB08668.1
2,5-Difluorphenylhydraziniumchlorid
2,5-Difluorphenylhydraziniumchlorid
Formel: C₆H₇ClF₂N₂
Molecular Weight: 180,58 g/mol
MDL: MFCD00013385
CAS-Nummer: 175135-73-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR